Order Entry
Ireland
ContactUsLinkComponent
VWR®, Pasteur Pipettes, Disposable
VWR®, Pasteur Pipettes, Disposable
Catalog # VWRU14673-010
Supplier:  VWR Collection
undefined
VWR®, Pasteur Pipettes, Disposable
Catalog # VWRU14673-010
Supplier:  VWR Collection

Specifications

  • Packed:
    200/box, 5 boxes/case
  • Capacity:
    2,0 ml
  • Description:
    Disposable pasteur pipette, short tip, without plug
  • Length:
    146 mm
  • Seq:
    01
  • Pk:
    1.000

Specifications

About this item

Borosilicate glass. These disposable pipettes feature fire-polished tops and a straight-cut tip.

  • Smooth, fire-polished tops facilitate bulb attachment
  • Pipettes have an inside diameter of 7 mm, which tapers to a straight-cut tip
  • Positive constriction located 25 mm from the top of the pipette facilitates plugging

Ideal for use in all general laboratory applications, including blood banks, hematology, chemistry, and bacteriology.