Anti-ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB Rabbit Polyclonal Antibody
Catalog # BOSSBS-5984R
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Abl-Interactor 2
- Antigen symbol:ABI2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:10152
- Antigen synonyms:argBPIA|ABI2B|AIP-1|ABI-2|argBPIB|AblBP3|SSH3BP2|argBP1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
ABI2 (Abelson interactor 2) belongs to the ABI family. The Abelson interactors are among a group of proteins that interact with Abl tyrosine kinase, a non-receptor tyrosine kinase whose activation results in the regulation of cytoskeleton function, cell growth and apoptosis in response to a variety of biological stimuli.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat