Anti-GRP75 Rabbit Polyclonal Antibody
BOSSBS-1469R
: Bioss
- Antibody type:Primary
- Antigen name:Heat shock 70kDa protein 9
- Antigen symbol:HSPA9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:3313
- Antigen synonyms:MOT|GRP-75|CRP40|GRP75|MOT2|HSPA9B|CSA|HEL-S-124m|PBP74|MTHSP75
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Immunogen:630-679/679
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Implicated in the control of cell proliferation and cellular aging. May also act as a chaperone.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: GRP75
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat