Order Entry
Ireland
ContactUsLinkComponent
Polytetrahydrofuran, M.W. 1000 stabilised for synthesis, Sigma-Aldrich®
Polytetrahydrofuran, M.W. 1000 stabilised for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Polytetrahydrofuran, M.W. 1000 stabilised for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
     - 10 g/l (20 °C)
  • Melting Point:
    23 - 28 °C
  • Boiling Point:
    250 °C
  • Vapor Pressure:
     - 0.1 hPa (20 °C)
  • Flash Point:
    240 °C
  • Refractive Index:
    1.466 (20 °C, 589 nm)
  • Density:
    0.982 g/cm3 (30 °C)
  • pH:
    7 (H2O, 20 °C) Aqueous solution
  • Average molecular mass:
    975 - 1025
  • Hydroxyl value:
    109 - 115
  • Storage temperature:
    +15...+25 °C

Specification Test Results

Related Information

Frequently Bought Together