Anti-6x His Mouse Monoclonal Antibody [clone: AD1.1.10]
Catalog # AGRIAS152937
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:His Tag
- Antigen symbol:6xHis
- Clonality:Monoclonal
- Clone:AD1.1.10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Reactivity:Zea mays,A. thaliana,H. vulgare
- Immunogen:Polyhistidine (HHHHHH) tagged polypeptide
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is not conjugated to any dye or enzyme. ,Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is not conjugated to any dye or enzyme.
Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: AD1.1.10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: A. thaliana, H. vulgare, Zea mays