Order Entry
Ireland
ContactUsLinkComponent
Anti-6x His Mouse Monoclonal Antibody (SureLight® R-Phycoerythrin) [clone: AD1.1.10]
Anti-6x His Mouse Monoclonal Antibody (SureLight® R-Phycoerythrin) [clone: AD1.1.10]
Catalog # AGRIAS152931
Supplier:  Agrisera
Anti-6x His Mouse Monoclonal Antibody (SureLight® R-Phycoerythrin) [clone: AD1.1.10]
Catalog # AGRIAS152931
Supplier:  Agrisera

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    His Tag
  • Antigen symbol:
    6xHis
  • Clonality:
    Monoclonal
  • Clone:
    AD1.1.10
  • Conjugation:
    SureLight® RPE (R-Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Reactivity:
    A. thaliana
  • Immunogen:
    Polyhistidine (HHHHHH) tagged polypeptide
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is directly conjugated to SureLight® R-Phycoerythrin. This is of advantage to shorten assay time by no need to use a secondary antibody to 6x Histidine tag.,Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is directly conjugated to SureLight® R-Phycoerythrin. This is of advantage to shorten assay time by no need to use a secondary antibody to 6x Histidine tag.

Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: AD1.1.10
Conjugation: SureLight® RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: A. thaliana