Anti-6x His Mouse Monoclonal Antibody (DyLight® 488) [clone: AD1.1.10]
Catalog # AGRIAS152928
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen name:His Tag
- Antigen symbol:6xHis
- Clonality:Monoclonal
- Clone:AD1.1.10
- Conjugation:DyLight® 488
- Flow cytometry:Yes
- Host:Mouse
- Reactivity:A. thaliana
- Immunogen:Polyhistidine (HHHHHH) tagged polypeptide
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is directly conjugated to DyLight® 488. This is of advantage to shorten assay time by no need to use a secondary antibody to 6x Histidine tag. DyLight® is a registered trade mark of Thermofisher Inc., and its subsidaries.,Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is directly conjugated to DyLight® 488. This is of advantage to shorten assay time by no need to use a secondary antibody to 6x Histidine tag. DyLight® is a registered trade mark of Thermofisher Inc., and its subsidaries.
Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: AD1.1.10
Conjugation: DyLight® 488
Epitope:
Host: Mouse
Isotype:
Reactivity: A. thaliana