Order Entry
Ireland
ContactUsLinkComponent
 
Anti-LEA4-25 Rabbit Polyclonal Antibody
  AGRIAS132757
 :  Agrisera
undefined
Anti-LEA4-25 Rabbit Polyclonal Antibody
  AGRIAS132757
 :  Agrisera

 

  • Antibody type:
    Primary
  • Antigen symbol:
    LEA4-25
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Phaseolus vulgaris
  • Western blot:
    Yes
  • Immunogen:
    recombinant Phaseolus vulgaris PvLEA 4-1 sequence derived from Pv01G142000
  • Size:
    50 µl
  • Pk:
    50 µl

 

 

LEA4-25 (Group 4-late embryogenesis abundant protein PvLEA4-25) belongs to a group of very hydrophilic proteins, described over 25 years ago as accumulating during late stages of plant seed development. Found in vegetative plant tissues following exposure to environmental stress. Synonymes: Putative late embryogenesis abundant protein LEA.,LEA4-25 (Group 4-late embryogenesis abundant protein PvLEA4-25) belongs to a group of very hydrophilic proteins, described over 25 years ago as accumulating during late stages of plant seed development. Found in vegetative plant tissues following exposure to environmental stress. Synonymes: Putative late embryogenesis abundant protein LEA.

Type: Primary
Antigen: LEA4-25
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Anabaena variablis, Anacystis nidulans r2, Aphanathece halophytica, Fremyella diplosiphon, Griffithsia monilis, Mastigocladus laminosus, Nostoc sP., Synechococcus sP. pcc 7002, atcc 27264, pr6, P. aeruginuso, Porphyridium cruentum, Rhodosorus marinus, Spirulina platensis, Synechecoccus lividus, Synechococcus elongatus pcc 7942, Synechococcus leopolensis, Tolypothrix tenuis