Order Entry
Ireland
ContactUsLinkComponent
pH meters, bench, sensION™+ pH31
pH meters, bench, sensION™+ pH31
Supplier:  Hach
Certificates
undefined
pH meters, bench, sensION™+ pH31
Supplier:  Hach

Specifications

  • Catalog No:
  • HACHLPV2115T98002
  • HACHLPV2121T98002
  • HACHLPV2111T98002
  • Model:
  • PH31
  • PH31
  • PH31
  • pH range:
  • −2...+19
  • −2...+19
  • −2...+19
  • pH resolution:
  • 0,1/0,01/0,001 (selectable)
  • 0,1/0,01/0,001 (selectable)
  • 0,1/0,01/0,001 (selectable)
  • pH accuracy:
  • ± 0,001
  • ± 0,001
  • ± 0,001
  • pH calibration:
  • 1-/2-/3-point (Automatic buffer recognition: technical DIN 19267, DIN 19266 and up to 3 custom buffers chosen by the user)
  • 1-/2-/3-point (Automatic buffer recognition: technical DIN 19267, DIN 19266 and up to 3 custom buffers chosen by the user)
  • 1-/2-/3-point (Automatic buffer recognition: technical DIN 19267, DIN 19266 and up to 3 custom buffers chosen by the user)
  • mV range:
  • ± 2000
  • ± 2000
  • ± 2000
  • mV resolution:
  • 0,1/1
  • 0,1/1
  • 0,1/1
  • mV accuracy:
  • ± 0,1
  • ± 0,1
  • ± 0,1
  • Temperature range:
  • −20...+150 °C
  • −20...+150 °C
  • −20...+150 °C
  • Temperature resolution:
  • 0,1 °C
  • 0,1 °C
  • 0,1 °C
  • Temperature accuracy:
  • ± 0,1 °C
  • ± 0,1 °C
  • ± 0,1 °C
  • Temperature compensation:
  • Automatic with Pt 1000, NTC 10KOhm temperature probe (ATC), or manual
  • Automatic with Pt 1000, NTC 10KOhm temperature probe (ATC), or manual
  • Automatic with Pt 1000, NTC 10KOhm temperature probe (ATC), or manual
  • Display:
  • Graphic, backlit liquid crystal, 128 × 64 dots
  • Graphic, backlit liquid crystal, 128 × 64 dots
  • Graphic, backlit liquid crystal, 128 × 64 dots
  • Power supply:
  • External power supply (100 - 240 V, 47 - 63 Hz)
  • External power supply (100 - 240 V, 47 - 63 Hz)
  • External power supply (100 - 240 V, 47 - 63 Hz)
  • Data logging capacity:
  • 330 measurements
  • 330 measurements
  • 330 measurements
  • Outputs:
  • RS 232 C
  • RS 232 C
  • RS 232 C
  • W×D×H:
  • 350×200×110 mm
  • 350×200×110 mm
  • 350×200×110 mm
  • Weight:
  • 1,1 kg
  • 1,1 kg
  • 1,1 kg

Specifications

Frequently Bought Together