Order Entry
Ireland
ContactUsLinkComponent
Anti-Foxp3 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: 3G3]
Anti-Foxp3 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: 3G3]
Supplier:  Tonbo Biosciences
Certificates
Anti-Foxp3 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: 3G3]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 60-5773-U100
  • 60-5773-U025
  • Antigen symbol:
  • FOXP3
  • FOXP3
  • Antigen name:
  • Forkhead Box P3
  • Forkhead Box P3
  • Antigen synonyms:
  • AIID|PIDX|JM2|DIETER|XPID|Scurfin|IPEX
  • AIID|PIDX|JM2|DIETER|XPID|Scurfin|IPEX
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)-Cy7®
  • PE (Phycoerythrin)-Cy7®
  • Clone:
  • 3G3
  • 3G3
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 50943/20371
  • 50943/20371
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • No
  • No
  • Storage temperature:
  • Store undiluted at 2-8 °C, protect from light, do not freeze.
  • Store undiluted at 2-8 °C, protect from light, do not freeze.

Specifications

Related Information

Frequently Bought Together