Order Entry
Ireland
ContactUsLinkComponent
VWR®, Autoclavable Infectious Waste Bag, High Strength
VWR®, Autoclavable Infectious Waste Bag, High Strength
Catalog # 129-0222
Supplier:  VWR Collection
undefined
VWR®, Autoclavable Infectious Waste Bag, High Strength
Catalog # 129-0222
Supplier:  VWR Collection

Specifications

  • Capacity:
  • Description:
    Red, with printed sign and temperature indicator
  • L×W:
    305×203 mm
  • Material:
    PP
  • Seq:
    03
  • Thickness:
    50 µm
  • Pk:
    400

Specifications

About this item

These high strength bags, made from PP, are ideal for hazardous waste collection and decontamination in an autoclave. Bags are formulated to withstand stress at the side seams and are strong enough to hold items in the laboratory. They are red and printed with a black biohazard symbol: 'Infectious Waste' and feature a temperature indicator patch.

  • Designed to handle dangerous materials
  • High-strength
  • Autoclavable to 135 °C (275 °F)
Certifications: Meets 480 g tear test (ASTM D1299-94A).
Delivery: Supplied with 12,7 cm twist ties.