Specifications
- Antibody type:Primary
- Antigen name:death-domain associated protein
- Antigen symbol:DAXX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Gene ID:1616
- Antigen synonyms:BING2|EAP1|DAP6
- UniProtKB:Q9UER7
- Storage temperature:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogen:E.coli-derived human Daxx recombinant protein (Position: K56-R345). Human Daxx shares 88.3% and 88.6% amino acid (aa) sequence identity with mouse and rat Daxx, respectively.
- Purification:Immunogen affinity purified.
- Size:100 μg / vial
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Death domain-associated protein 6(DAXX) detection. Tested with WB, IHC-P, ICC in Human;Rat.
DAXX (death-domain associated protein) also known as DAP6 (Death-associated protein 6) or BING2, was first discovered through its cytoplasmic interaction with the classical death receptor Fas. Human DAXX encodes a 740-amino acid polypeptide containing a nuclear localization signal. Functional analyses by Yang et al. (1997) demonstrated that Daxx binds to the Fas death domain and enhances Fas-mediated apoptosis. The authors suggested that DAXX and FADD define 2 distinct apoptotic pathways downstream of Fas. The DAXX gene is mapped to human chromosome 6p21.3 by somatic cell hybrid panels and fluorescence in situ hybridization, a region containing the HLA and putative autoimmune disease genes. MSP58 overexpression relieved DAXX-mediated transcriptional repression. Immunoprecipitation and Western blot analysis with DAXX mutants showed that the N terminus of DAXX interacts with the C terminus of DMAP. Transient expression of DAXX or DMAP1 caused repression of glucocorticoid receptor-mediated transcription.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: Daxx
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat