Anti-IFNK Antibody Pair
ABNOH00056832-AP51
: Abnova
- Antigen name:Interferon kappa
- Antigen symbol:IFNK
- Host:Multiple
- Reactivity:Human
- Gene ID:56832
- Antigen synonyms:IFNT1|INFE1
- Storage temperature:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Size:1 Set
- Pk:1 SET
This Set comes with matched Antibody Pair to detect and quantify protein level of human IFNK.
Antibody pair set content: 1. Capture Antibody: Mouse Monoclonal Anti-IFNK (100 µg); 2. Detection Antibody: Rabbit purified Polyclonal Anti-IFNK (50 µg);
Type:
Antigen: IFNK
Clonality:
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human