Megabase DNA standards
Catalog # LONZ50411
Supplier: LONZA
CAS Number:
Specifications
- Pk:5
- Description:Saccharomyces cerevisiae DNA standards, 220 kb - 1 Mb, 5 plugs, (10 ± 2 μg DNA/plug)
- Seq:02
Specifications
About this item
Megabase DNA standards are specially prepared in InCert™ Agarose for use in pulsed field gel electrophoresis (PFGE).
- Minimal lot-to-lot variation
- Each lot is performance tested for reliable results