Accessories for plasma 40/plasma 400
Catalog # PERCN0690676
Supplier: PerkinElmer
Specifications
- Description:GemTip cross-flow II tip kits
- Pk:1
Specifications
About this item
This tip set includes a clear sapphire sample tip (0,3 mm Int. Ø) and a red ruby argon tip (0,22 mm Int. Ø).
The tips are made of PEEK with the gem inserts noted for maximum corrosion resistance.