Order Entry
Ireland
ContactUsLinkComponent
Hanks balanced salt solution (HBSS)
Hanks balanced salt solution (HBSS)
Catalog # LONZ04-315Q
Supplier:  LONZA
CAS Number:  
Hanks balanced salt solution (HBSS)
Catalog # LONZ04-315Q
Supplier:  LONZA
CAS Number:  
Specifications
  • Pk:
    1 L
  • Description:
    BSS, without Ca++, Mg++ or phenol red
  • Seq:
    05
Specifications
About this item
Hanks balanced salt solution is used in a wide variety of tissue culture applications like to maintain a physiological pH for cells maintained in non CO₂ conditions, diluting cells before counting, washing cells before dissociating and transporting cells or tissues.