Order Entry
Ireland
ContactUsLinkComponent
 
Anti-Adiponectin Chicken Polyclonal Antibody
  BSENC-1524-100
 :  Biosensis
undefined
Anti-Adiponectin Chicken Polyclonal Antibody
  BSENC-1524-100
 :  Biosensis

 

  • Antibody type:
    Primary
  • Antigen name:
    Adiponectin
  • Antigen symbol:
    ADIPOQ
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Chicken
  • Isotype:
    IgY
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    Maps to amino acids 230-244 of Human Adiponectin protein
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Immunogen:
    A synthetic peptide from human Adiponectin (230-244 aa).
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Adiponectin is synthesized by adipocytes and is involved in the control of fat metabolism and insulin sensitivity, with direct anti-diabetic, anti-atherogenic and anti-inflammatory activities (ref: SWISSPROT).

This antibody detects adiponectin protein at approx 30 kDa in western blots from human, mouse and rat samples. Plasma makes a good positive control. The antibody has not been tested in IHC
Application Information:
WB. Suggested dilution of 1:2,000-1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Adiponectin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Maps to amino acids 230-244 of Human Adiponectin protein
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat