Anti-GST Mouse Monoclonal Antibody [clone: BGSTB6]
Catalog # BSENM-1306-100
Supplier: Biosensis
Specifications
- Antibody type:Tag
- Antigen name:Glutathione-S-Transferase
- Antigen symbol:GST
- Clonality:Monoclonal
- Clone:BGSTB6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Epitope:GST tag
- Cross adsorption:No
- Format:Liquid
- Immunogen:Mouse monoclonal anti-GST tag antibody was produced by immunizing mice with glutathione S-transferase (GST) peptide.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
GST (Glutathione S-Transferase) is a 26 kDa protein encoded by the Schistosoma japonicum.
Mouse monoclonal anti-GST tag antibody detects over expressed glutathione- transferase (GST) fusion proteins.
Application Information:
Western Blotting (WB), Immunohistochemistry (IH) and Immunoprecipitation (IP). Suggested dilutions for WB of 1:1000, IS and IP at 1:200. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Tag
Antigen: GST
Clonality: Monoclonal
Clone: BGSTB6
Conjugation: Unconjugated
Epitope: GST tag
Host: Mouse
Isotype:
Reactivity: