Specifications
- Antibody type:Tag
- Antigen name:DYKDDDDK tag
- Antigen symbol:DYKDDDDK
- Clonality:Monoclonal
- Clone:BFtag01
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Epitope:DYKDDDDK
- Cross adsorption:No
- Format:Liquid
- Immunogen:A synthetic peptide (DYKDDDDK) coupled to KLH.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
The D-tag system utilises a short hydrophilic peptide (DYKDDDDK) that is fused to either the N- or C-terminus of the protein of interest. It can be used in conjunction with other tags such as the 6X His tag.
Mouse monoclonal anti D-tag antibody recognises over-expressed proteins containing the D-tag fused to either amino- or carboxy-termini of targeted proteins in transfected mammalian cells.
Application Information:
Western Blotting (WB), Immunohistochemistry (IH) and Immunoprecipitation (IP). Suggested dilutions: WB at 1:100, IS and IP at 1:20. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Tag
Antigen: DYKDDDDK
Clonality: Monoclonal
Clone: BFtag01
Conjugation: Unconjugated
Epitope: DYKDDDDK
Host: Mouse
Isotype:
Reactivity: