Nessler colour comparison tubes, KIMBLE®
SCERSP45310-50100
- Description:Nessler Color Comparison Tubes
- Length:200 mm
- Seq:03
- Graduation:50 - 100 ml
- Pk:6
- Scale length:145-158 mm
Manufactured from 33 expansion, low extractable borosilicate glass.
- Shadowless bottom of each tube transmits undistorted light
- Rings and legend are durable white ceramic enamel
- Matched or unmatched available
These Nessler tubes are suitable for colour comparison.
: Glass conforms to USP Type I and ASTM E438 Type I Class A requirements.