Sleeve protectors, PE, Hopen®
Supplier: Medicom
Certificates
About this item
These disposable sleeve protectors made of PE protect the arms against dirt.
- Elasticated
- Latex-free
- Available in two different colours: White and blue
Ideal for use in non hazardous applications, such as cleaning, maintenance and food processing.
Thickness: 20 µm
Specifications
- Catalog No:
- HOPE20.300
- HOPE20.100
- Garment application:
- Laboratory, Food
- Laboratory, Food