Overalls, Hopen®
: Medicom
These disposable overalls with collar are made of breathable PP.
- Lightweight polypropylene is highly breathable
- With collar and elasticated cuffs and ankles
- With zip closure (without flap)
Ideal for use in the chemical and petrochemical industry, for food applications, cleaning and non hazardous industrial applications.
Weight: 40 g/m²
- Catalog No:
- HOPE40.101/05
- HOPE40.101/02
- HOPE40.101/04
- HOPE40.101/06
- HOPE40.101/03
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Garment application:
- Laboratory, Food
- Laboratory, Food
- Laboratory, Food
- Laboratory, Food
- Laboratory, Food
- Weight:
- 40 g/m²
- 40 g/m²
- 40 g/m²
- 40 g/m²
- 40 g/m²
Frequently Bought Together