Sample processing manifolds, VacMaster™ 10 and 20
Catalog # ARGO121-2012
Supplier: Argonaut Technologies
Specifications
- Description:Sample processing manifold, VacMaster™ 20, complete with 12 mm Ø collection tube rack
- Seq:05
- Environmentally Preferable:
- Pk:1
Specifications
About this item
These robust manifolds are constructed from high quality, inert materials (glass, PTFE and PE), eliminating sample contamination and allowing all common solvents and buffers to be used. System allows up to 10 (VacMaster™ 10) or 20 (VacMaster™ 20) SPE or filtration columns to be processed simultaneously under vacuum. Unused ports are sealed when not required. Easy to use vacuum controls provide excellent sample flow control, leading to reproducible, reliable extraction conditions.
- Disposable stainless steel needles fitted as standard
- Clear view of sample processing operation
- Vacuum safety valve fitted as standard
Delivery: Both systems come complete with glass tank fitted with PTFE waste outlet and drain tube PTFE lid complete with 10 or 20 PTFE needle retainers, stainless steel needles and port sealing plugs. Lid also houses vacuum control arm including on/off valve, fine vacuum control and vacuum release valve. Complete collection tube rack (PE) with 10, 12 or 16 mm collection tube positions.