Order Entry
Ireland
ContactUsLinkComponent
Anti-CACNA1G Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S178A-9]
Anti-CACNA1G Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S178A-9]
Catalog # ABNOMAB11564
Supplier:  Abnova
Anti-CACNA1G Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: S178A-9]
Catalog # ABNOMAB11564
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Calcium Channel, Voltage-dependent, T type, alpha 1G subunit
  • Antigen symbol:
    CACNA1G
  • Clonality:
    Monoclonal
  • Clone:
    S178A-9
  • Conjugation:
    RPE (R-Phycoerythrin)
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    12291
  • Antigen synonyms:
    Cav3.1d|[a]1G|mKIAA1123|a1G|alpha-1G
  • Amino acid number:
    2052 to 2172
  • Storage buffer:
    PBS, pH 7,4 (50% glycerol)
  • Storage temperature:
    Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant fusion protien corresponding to amino acids 2052-2172 at C-terminus of mouse Cacna1g.
  • Purification:
    Protein G Purified
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against recombinant of Cacna1g.

Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: CACNA1G
Clonality: Monoclonal
Clone: S178A-9
Conjugation: RPE (R-Phycoerythrin)
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: