Anti-HMOX2 Mouse Monoclonal Antibody [clone: S3]
Catalog # ABNOH00003163-M01A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Heme Oxidase 2
- Antigen symbol:HMOX2
- Clonality:Monoclonal
- Clone:S3
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1 kappa
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites fluid
- Gene ID:3163
- Antigen synonyms:HO-2
- Amino acid number:1 to 316
- Sequence:MSAEVETSEGVDESEKKNSGALEKENQMRMADLSELLKEGTKEAHDRAENTQFVKDFLKGNIKKELFKLATTALYFTYSALEEEMERNKDHPAFAPLYFPMELHRKEALTKDMEYFFGENWEEQVQCPKAAQKYVERIHYIGQNEPELLVAHAYTRYMGDLSGGQVLKKVAQRALKLPSTGEGTQFYLFENVDNAQQFKQLYRARMNALDLNMKTKERIVEEANKAFEYNMQIFNELDQAGSTLARETLEDGFPVHDGKGDMRKCPFYAAEQDKGALEGSSCPFRTAMAVLRKPSLQFILAAGVALAAGLLAWYYM
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:HMOX2 (AAH02396.1, 1 a.a. ~ 316 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:200 μl
- Pk:200 µg
Specifications
About this item
Mouse monoclonal antibody raised against a full-length recombinant HMOX2.
Type: Primary
Antigen: HMOX2
Clonality: Monoclonal
Clone: S3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: