Anti-PTPRC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: BRA-11]
Catalog # ABNOMAB12144
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Monoclonal
- Clone:BRA-11
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Cross adsorption:No
- Format:Liquid
- Gene ID:5788
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Storage buffer:PBS (0,05% BSA, 0,05% sodium azide).
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Non-T, non-B CALLA positive, ALL cell line REH.
- Purification:Protein A/G purification
- Pk:500 µl
Specifications
About this item
Mouse monoclonal antibody raised against human PTPRC.
Recommended Dilutions: Flow Cytometry: 2.5 ?l/million cells; Immunohistochemistry
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: BRA-11
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: