Anti-CXCL2 Mouse Monoclonal Antibody [clone: 2D10]
Catalog # ABNOH00002920-M21
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Chemokine (C-X-C motif) Ligand 2
- Antigen symbol:CXCL2
- Clonality:Monoclonal
- Clone:2D10
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a kappa
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:2920
- Antigen synonyms:MIP-2|CINC-2a|GROb|MGSA-b|MIP-2a|GRO2|SCYB2|CINC-3|MIP2|GRO beta|MIP2A
- Amino acid number:1 to 100
- Storage buffer:1x PBS, pH 7,4
- Sequence:MARATLSAAPSNPRLLRVALLLLLLVAASRRAAGAPLATELRCQCLQTLQGIHLKNIQSVKVKSPGPHCAQTEVIATLKNGQKACLNPASPMVKKIIEKM
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:CXCL2 (NP_002080.1, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against a partial recombinant CXCL2.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: CXCL2
Clonality: Monoclonal
Clone: 2D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: