Order Entry
Ireland
ContactUsLinkComponent
Anti-TP53 Mouse Monoclonal Antibody [clone: SPM589]
Anti-TP53 Mouse Monoclonal Antibody [clone: SPM589]
Catalog # ABNOMAB12059
Supplier:  Abnova
Anti-TP53 Mouse Monoclonal Antibody [clone: SPM589]
Catalog # ABNOMAB12059
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tumor protein p53
  • Antigen symbol:
    P53
  • Clonality:
    Monoclonal
  • Clone:
    SPM589
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    7157
  • Antigen synonyms:
    Tumor suppressor p53|Cellular tumor antigen p53|TP53|Antigen NY-CO-13|Phosphoprotein p53
  • Storage buffer:
    PBS (0,05% BSA, 0,05% sodium azide).
  • Storage temperature:
    Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human TP53.
  • Purification:
    Protein A/G purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against recombinant TP53.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: TP53
Clonality: Monoclonal
Clone: SPM589
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: