Order Entry
Ireland
ContactUsLinkComponent
Anti-POMC Mouse Monoclonal Antibody [clone: SPM333]
Anti-POMC Mouse Monoclonal Antibody [clone: SPM333]
  ABNOMAB12048
 :  Abnova
Anti-POMC Mouse Monoclonal Antibody [clone: SPM333]
  ABNOMAB12048
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Proopiomelanocortin
  • Antigen symbol:
    POMC
  • Clonality:
    Monoclonal
  • Clone:
    SPM333
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    5443
  • Antigen synonyms:
    OTTHUMP00000119991|adrenoCorticotropin/ beta-lipotropin/ alpha-melanocyte stimulating hormone/ beta-melanocyte stimulating|NPP|beta-LPH|lipotropin beta|hormone/ beta-endorphin|alpha-melanocyte-stimulating hormone|adrenocorticotropic hormone|lipotropin gamm|POC|beta-melanocyte-stimulating hormone|gamma-MSH|alpha-MSH|beta-endorphin|CLIP|LPH|Precursor of MSH|adrenoCorticotropin|POMC|proopiomelanocortin|gamma-LPH|OTTHUMP00000200964|Corticotropin-like intermediary peptide|ACTH
  • Storage buffer:
    PBS (0,05% BSA, 0,05% sodium azide).
  • Storage temperature:
    Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to amina acid 1-24 of human POMC.
  • Purification:
    Protein A/G purification
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against synthetic peptide of POMC.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: POMC
Clonality: Monoclonal
Clone: SPM333
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: