Anti-SLC5A5 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: FP5]
Catalog # ABNOMAB11510
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 5 (low affinity glucose cotransporter) Member 5
- Antigen symbol:Solute carrier family 5, member 5
- Clonality:Monoclonal
- Clone:FP5
- Conjugation:RPE (R-Phycoerythrin)
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:6528
- Antigen synonyms:TDH1|NIS|Sodium-Iodide Symporter|CNGA1/3
- Amino acid number:468 to 643
- Storage buffer:PBS, pH 7,4 (50% glycerol)
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant mannose fusion protien corresponding to amino acids 468-643 of human SLC5A5.
- Purification:Protein G purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant of SLC5A5.
Recommended Dilutions: Immunofluorescence Immunohistochemistry Western Blot: 1:1000
Type: Primary
Antigen: SLC5A5
Clonality: Monoclonal
Clone: FP5
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: