Order Entry
Ireland
ContactUsLinkComponent
Anti-SLC5A5 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: FP5]
Anti-SLC5A5 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: FP5]
  ABNOMAB11509
 :  Abnova
Anti-SLC5A5 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: FP5]
  ABNOMAB11509
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Solute Carrier Family 5 (low affinity glucose cotransporter) Member 5
  • Antigen symbol:
    Solute carrier family 5, member 5
  • Clonality:
    Monoclonal
  • Clone:
    FP5
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    6528
  • Antigen synonyms:
    TDH1|NIS|Sodium-Iodide Symporter|CNGA1/3
  • Amino acid number:
    468 to 643
  • Storage buffer:
    PBS, pH 7,4 (50% glycerol)
  • Storage temperature:
    Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant mannose fusion protien corresponding to amino acids 468-643 of human SLC5A5.
  • Purification:
    Protein G purification
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against recombinant of SLC5A5.

Recommended Dilutions: Immunofluorescence Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: SLC5A5
Clonality: Monoclonal
Clone: FP5
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: