Anti-Clostridium difficile Toxin B Chicken Polyclonal Antibody
Catalog # ABNOPAB29154
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Clostridium difficile Toxin B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:IgY
- Cross adsorption:No
- Format:Liquid
- Storage buffer:PBS, pH 7,2 (0,075% sodium azide).
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified Clostridium difficile Toxin B with Freund's adjuvant.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Chicken polyclonal antibody raised against Clostridium difficile Toxin B.
Type: Primary
Antigen: Clostridium difficile Toxin B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: