Order Entry
Ireland
ContactUsLinkComponent
Anti-MAGEA1 Mouse Monoclonal Antibody [clone: SPM282]
Anti-MAGEA1 Mouse Monoclonal Antibody [clone: SPM282]
Catalog # ABNOMAB12108
Supplier:  Abnova
Anti-MAGEA1 Mouse Monoclonal Antibody [clone: SPM282]
Catalog # ABNOMAB12108
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Melanoma-Associated Antigen 1
  • Antigen symbol:
    MAGEA1
  • Clonality:
    Monoclonal
  • Clone:
    SPM282
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    4100
  • Antigen synonyms:
    melanoma antigen family A 1|antigen MZ2-E|MAGE-1 antigen|cancer/testis antigen family 1|member 1|melanoma-associated antigen 1|melanoma antigen MAGE-1|melanoma-associated antigen MZ2-E|cancer/testis antigen 1.1
  • Storage buffer:
    PBS (0,05% BSA, 0,05% sodium azide).
  • Storage temperature:
    Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human full length MAGEA1.
  • Purification:
    Protein A/G purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against MAGEA1.

Recommended Dilutions: Immunohistochemistry: 0.5-1 ?g/ml; Western Blot: 0.5-1 ?g/ml

Type: Primary
Antigen: MAGEA1
Clonality: Monoclonal
Clone: SPM282
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: