Anti-APP Rabbit Polyclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex))
ABNOPAB28925
: Abnova
- Antigen name:Amyloid beta (A4) Precursor Protein
- Antigen symbol:APP
- Clonality:Polyclonal
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:351
- Antigen synonyms:CTFgamma|CVAP|ABETA|APPI|AD1|PN-II|ABPP|AAA|PN2
- Storage buffer:PBS pH 7,4, 50% glycerol and 0,09% sodium azide
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Fibrils prepared from human Abeta42 peptide.
- Tested applications:Dot Blot
- Purification:Protein A affinity purification
- Pk:100 µl
Rabbit polyclonal antibody against amyloid fibrils and fibrillar oligomers.
Recommended Dilutions: Dot Blot: 1:1000
Type:
Antigen: APP
Clonality: Polyclonal
Clone:
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Rabbit
Isotype:
Reactivity: