Anti-OBSCN Mouse Monoclonal Antibody [clone: 5C20]
Catalog # ABNOH00084033-M02A
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Obscurin, cytoskeletal calmodulin And titin-interacting RhoGEF
- Antigen symbol:OBSCN
- Clonality:Monoclonal
- Clone:5C20
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a kappa
- Cross adsorption:No
- Format:Liquid
- Form:Ascites fluid
- Gene ID:84033
- Antigen synonyms:ARHGEF30, UNC89
- Amino acid number:1551 to 1649
- Sequence:KAGMGPYSSPSEQVLLGGPSHLASEEESQGRSAQPLPSTKTFAFQTQIQRGRFSVVRQCWEKASGRALAAKIIPYHPKDKTAVLREYEALKGLRHPHLA
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:OBSCN (XP_290923, 1551 a.a. ~ 1649 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:200 μl
- Pk:200 µg
Specifications
About this item
Mouse monoclonal antibody raised against a partial recombinant OBSCN.
Type: Primary
Antigen: OBSCN
Clonality: Monoclonal
Clone: 5C20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: