Order Entry
Ireland
ContactUsLinkComponent
Anti-NME1 Mouse Monoclonal Antibody [clone: 4B2]
Anti-NME1 Mouse Monoclonal Antibody [clone: 4B2]
  ABNOMAB10833
 :  Abnova
Anti-NME1 Mouse Monoclonal Antibody [clone: 4B2]
  ABNOMAB10833
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    NME/NM23 nucleoside diphosphate kinase 1
  • Antigen symbol:
    NME1
  • Clonality:
    Monoclonal
  • Clone:
    4B2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Ascites
  • Gene ID:
    4830
  • Antigen synonyms:
    NM23|NDPKA|NDKA|NBS|AWD|GAAD|NM23-H1|NDPK-A|NB
  • Storage buffer:
    0,03% sodium azide
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to human NME1.
  • Size:
    100 μl
  • Pk:
    100 µl

 

 

Mouse monoclonal antibody raised against partial recombinant NME1.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400

Type: Primary
Antigen: NME1
Clonality: Monoclonal
Clone: 4B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human