Anti-HFE Mouse Monoclonal Antibody [clone: 3F1]
ABNOMAB10680
: Abnova
- Antibody type:Primary
- Antigen name:Hemochromatosis
- Antigen symbol:HFE
- Clonality:Monoclonal
- Clone:3F1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:3077
- Antigen synonyms:TFQTL2|hereditary hemochromatosis protein HLA-H|hereditary hemochromatosis protein|MVCD7|MHC class I-like protein HFE|HLA-H|HFE1|high Fe|HH
- Storage buffer:0,03% sodium azide
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human HFE.
- Size:100 μl
- Pk:100 µl
Mouse monoclonal antibody raised against partial recombinant HFE.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000
Type: Primary
Antigen: HFE
Clonality: Monoclonal
Clone: 3F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human