Anti-CK6B Mouse Monoclonal Antibody [clone: 1C12]
ABNOH00003854-M04
: Abnova
- Antibody type:Primary
- Antigen name:Cytokeratin 6B
- Antigen symbol:CK6B
- Clonality:Monoclonal
- Clone:1C12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3854
- Antigen synonyms:PC2|K6B|CK-6B|PC4|CK6B|KRTL1
- Amino acid number:1 to 564
- Storage buffer:1x PBS, pH 7,4
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:KRT6B (AAH34535, 1 a.a. ~ 564 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:100 µg
- Pk:100 µg
Mouse monoclonal antibody raised against a full-length recombinant KRT6B.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: CK6B
Clonality: Monoclonal
Clone: 1C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human