Order Entry
Ireland
ContactUsLinkComponent
Anti-SLC7A5 Mouse Monoclonal Antibody [clone: 4D9]
Anti-SLC7A5 Mouse Monoclonal Antibody [clone: 4D9]
Catalog # ABNOMAB1711
Supplier:  Abnova
Anti-SLC7A5 Mouse Monoclonal Antibody [clone: 4D9]
Catalog # ABNOMAB1711
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 7 (amino acid transporter light chain, L system), member 5
  • Antigen symbol:
    Solute carrier family 7, member 5
  • Clonality:
    Monoclonal
  • Clone:
    4D9
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    8140
  • Antigen synonyms:
    E16|4F2LC|hLAT1|LAT1|CD98|MPE16|D16S469E
  • Storage buffer:
    PBS (0,1% BSA, 0,1% proclin)
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to human SLC7A5.
  • Size:
    20 μg
  • Pk:
    20 µg

Specifications

About this item

Mouse monoclonal antibody raised against synthetic peptide of SLC7A5.

Recommended Dilutions: Immunohistochemistry: 2-5 ?g/ml

Type: Primary
Antigen: SLC7A5
Clonality: Monoclonal
Clone: 4D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human