Anti-LPAR2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB10124
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Lysophosphatidic acid receptor 2
- Antigen symbol:LPAR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:9170
- Antigen synonyms:LPA-2|EDG4|LPA2|EDG-4
- Storage buffer:PBS (0,08% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to N-terminus of human LPAR2.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of LPAR2.
Recommended Dilutions: Western Blot: 5 to 10 ?g/ml
Type: Primary
Antigen: LPAR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human