Anti-SLC16A7 Rabbit Polyclonal Antibody
Catalog # ABNOPAB10132
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 16 Member A7
- Antigen symbol:Solute carrier family 16, member 7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:9194
- Antigen synonyms:MCT2
- Storage buffer:PBS (0,08% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human SLC16A7.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of SLC16A7.
Recommended Dilutions: Western Blot: 1:400
Type: Primary
Antigen: SLC16A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human