Order Entry
Ireland
ContactUsLinkComponent
 
Anti-ATP1B1 Mouse Monoclonal Antibody [clone: 464.8 (also known as 8A)]
  ABNOMAB2323
 :  Abnova
undefined
Anti-ATP1B1 Mouse Monoclonal Antibody [clone: 464.8 (also known as 8A)]
  ABNOMAB2323
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    ATPase, Na+/K+ transporting, beta 1 Polypeptide
  • Antigen symbol:
    ATP1B1
  • Clonality:
    Monoclonal
  • Clone:
    464.8 (also known as 8A)
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Pig,
    Rabbit,
    Human,
    Bovine,
    Dog,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    100008749
  • Antigen synonyms:
    ATP1B1
  • Storage buffer:
    buffer containing 0,1% sodium azide
  • Storage temperature:
    Store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified rabbit ATP1B1.
  • Size:
    100 μl
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against native ATP1B1.

Recommended Dilutions: Western Blot: 1:2500-1:10000

Type: Primary
Antigen: ATP1B1
Clonality: Monoclonal
Clone: 464.8 (also known as 8A)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Bovine, Dog, Human, Pig, Rabbit, Rat