Anti-FANCD2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB12428
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fanconi Anemia Complementation Group D2
- Antigen symbol:FANCD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:2177
- Antigen synonyms:FAD|FA4|FANCD|FACD|FA-D2|FAD2
- Storage buffer:Tris-glycine, 150 mM NaCl (0,05% sodium azide)
- Storage temperature:Store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to N-terminus of human FANCD2.
- Size:50 μl
- Pk:50 µl
Specifications
About this item
Rabbit polyclonal antibody raised against partial recombinant FANCD2.
Recommended Dilutions: Immunofluorescence: 1:200-1:500; Western Blot: 1:10000-1:20000; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2.5-5 ?g/ml
Type: Primary
Antigen: FANCD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse