Order Entry
Ireland
ContactUsLinkComponent
Anti-EPB41L3 Rabbit Polyclonal Antibody
Anti-EPB41L3 Rabbit Polyclonal Antibody
Catalog # ABNOPAB12438
Supplier:  Abnova
Anti-EPB41L3 Rabbit Polyclonal Antibody
Catalog # ABNOPAB12438
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Erythrocyte membrane protein band 4.1-like 3
  • Antigen symbol:
    EPB41L3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Xenopus,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    23136
  • Antigen synonyms:
    differentially expressed in adenocarcinoma of the lung|EPB41L3|DAL-1|erythrocyte membrane protein band 4.1-like 3 |HGNC:3380|FLJ37633|DAL1|4.1B|KIAA0987
  • Amino acid number:
    400 to 500
  • Storage buffer:
    PBS (30% glycerol, 0,1% sodium azide)
  • Storage temperature:
    Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to amino acids 400-500 of human EPB41L3.
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic peptide of EPB41L3.

Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: EPB41L3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus