Anti-FXR2 Mouse Monoclonal Antibody [clone: A42]
ABNOMAB2493
: Abnova
- Antibody type:Primary
- Antigen name:fragile X mental retardation, autosomal homolog 2
- Antigen symbol:FXR2
- Clonality:Monoclonal
- Clone:A42
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:9513
- Antigen synonyms:FMR1L2|FXR2P
- Storage buffer:PBS (0,1% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant His fusion protein corresponding to human FXR2.
- Size:100 μl
- Pk:100 µl
Mouse monoclonal antibody raised against recombinant FXR2.
Type: Primary
Antigen: FXR2
Clonality: Monoclonal
Clone: A42
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human