Anti-ACSL1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB12497
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Acyl-CoA Synthetase Long-chain Family Member 1
- Antigen symbol:ACSL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Primates,Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:2180
- Antigen synonyms:FACL1|FACL2|LACS|LACS1|LACS2|ACS1
- Amino acid number:1 to 100
- Storage buffer:PBS (30% glycerol, 0,1% sodium azide)
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 1-100 of human ACSL1.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of ACSL1.
Recommended Dilutions: Western Blot: 1 ?g/ml
Type: Primary
Antigen: ACSL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Primates, Rat