Order Entry
Ireland
ContactUsLinkComponent
Anti-PRKAA1 Rabbit Polyclonal Antibody
Anti-PRKAA1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB12602
Supplier:  Abnova
undefined
Anti-PRKAA1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB12602
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase AMP-activated Catalytic Subunit Alpha 1
  • Antigen symbol:
    PRKAA1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Insect,
    Chicken,
    Human,
    Bovine,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    5562
  • Antigen synonyms:
    catalytic alpha-1 chain|catalytic|AMP-activated protein kinase|5'-AMP-activated protein kinase catalytic subunit alpha-1|AMPK|5'-AMP-activated protein kinase|tau-protein kinase PRKAA1|AMPK subunit alpha-1|AMPK alpha 1|AMP -activate kinase alpha 1 subunit|alpha-1|AMPKa1
  • Modification:
    phospho-Thr172
  • Storage buffer:
    TBS, pH 7,2 (10% Proclin300)
  • Sequence:
    LRTS
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Synthetic phosphopeptide corresponding to residues surrounding T172 of human PRKAA1.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Rabbit polyclonal antibody raised against synthetic phosphopeptide of PRKAA1.

Recommended Dilutions: Western Blot: 0.1-1 ?g/ml; ELISA: 0.01-0.1 ?g/ml; Immunoprecipitation: 2-5 ?g/ml

Type: Primary
Antigen: PRKAA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Dog, Human, Insect, Mouse, Rat