Anti-FCGR3B Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: MEM-154]
Catalog # ABNOMAB4541
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fc fragment of IgG, low affinity 3B, receptor (CD16b)
- Antigen symbol:FCGR3B
- Clonality:Monoclonal
- Clone:MEM-154
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:2215
- Antigen synonyms:CD16b|CD16|FCRIII|FCGR3|FCG3|FCR-10|FCRIIIb
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified FCGR3B from human granulocytes.
- Size:100 Reactions
- Pk:100 Tests
Specifications
About this item
Mouse monoclonal antibody raised against native FCGR3B.
Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: FCGR3B
Clonality: Monoclonal
Clone: MEM-154
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human