Anti-ITGB1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-101A]
ABNOMAB4545
: Abnova
- Antibody type:Primary
- Antigen name:Integrin Beta 1
- Antigen symbol:ITGB1
- Clonality:Monoclonal
- Clone:MEM-101A
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Pig,Human,Dog
- Cross adsorption:No
- Format:Liquid
- Gene ID:3688
- Antigen synonyms:FNRB|integrin VLA-4 beta subunit|VLA-BETA|VLAB|VLA-4 subunit beta|CD29|beta polypeptide|MDF2|MSK12|GPIIA|Fibronectin receptor subunit beta|very late activation protein
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified ITGB1 from Raji Burkitt's lymphoma cell line.
- Size:100 Reactions
- Pk:100 Tests
Mouse monoclonal antibody raised against native ITGB1.
Recommended Dilutions: Flow Cytometry: 10 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)
Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone: MEM-101A
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Pig