Anti-CD80 Mouse Monoclonal Antibody [clone: MEM-233]
Catalog # ABNOMAB3845
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:CD80 molecule
- Antigen symbol:CD80
- Clonality:Monoclonal
- Clone:MEM-233
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:941
- Antigen synonyms:TSA1|B71|Ly-53|MIC17|Cluster of Differentiation 80|Cd28l|Ly53
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant Fc fusion protein corresponding to extracellular domain of human CD80.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant CD80.
Type: Primary
Antigen: CD80
Clonality: Monoclonal
Clone: MEM-233
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human